Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2-6 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89119417 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-417 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-417 Supplier Abnova Corporation Supplier No. PAB17751
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NKX2-6.

Homeobox-containing genes play critical roles in regulating tissue-specific gene expression essential for tissue differentiation, as well as determining the temporal and spatial patterns of development (Shiojima et al., 1995 [PubMed 7665173]). It has been demonstrated that a Drosophila homeobox-containing gene called 'tinman' is essential for development of the heart-like dorsal vessel. NKX2-6 is a vertebrate homolog of 'tinman' (Newman and Krieg, 1998 [PubMed 9621430]).[supplied by OMIM

Specifications

Antigen NKX2-6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NKX2-6.
Dilution Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene NKX2-6
Gene Alias CSX2/NKX4-2
Gene Symbols NKX2-6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human NKX2-6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137814
Test Specificity This antibody detects endogenous levels of total NKX2-6 protein.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.