Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRC4 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114080 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-080 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-080 Supplier Abnova Corporation Supplier No. PAB0258
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NLRC4.

In C. elegans, Ced4 binds and activates Ced3, an apoptotic initiator caspase, via caspase-associated recruitment domains (CARDs). Human Ced4 homologs include APAF1 (MIM 602233), NOD1/CARD4 (MIM 605980), and NOD2/CARD15 (MIM 605956). These proteins have at least 1 N-terminal CARD domain followed by a centrally located nucleotide-binding domain (NBD or NACHT) and a C-terminal regulatory domain, found only in mammals, that contains either WD40 repeats or leucine-rich repeats (LRRs). CARD12 is a member of the Ced4 family and can induce apoptosis.[supplied by OMIM

Sequence: EEAPETYIPSRAVSLFFN

Specifications

Antigen NLRC4
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NLRC4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene NLRC4
Gene Alias CARD12/CLAN/CLAN1/CLANA/CLANB/CLANC/CLAND/CLR2.1/IPAF
Gene Symbols NLRC4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 637-654 of human NLRC4.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58484
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.