Learn More
NLRP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NLRP1.
Supplier: Abnova Corporation PAB0262
Description
This gene encodes a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq
Sequence: PSSPDHESPSQESPNAPTSTSpecifications
NLRP1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NLRP1. | |
In serum (0.05% sodium azide) | |
CARD7/CLR17.1/DEFCAP/DEFCAP-L/S/DKFZp586O1822/KIAA0926/NAC/NALP1/PP1044/SLEV1/VAMAS1 | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
NLRP1 | |
NLRP1 | |
A synthetic peptide corresponding to amino acids 161-180 of human NLRP1. | |
RUO | |
22861 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only