Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114083 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-083 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-083 Supplier Abnova Corporation Supplier No. PAB0261
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NLRP1.

This gene encodes a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq

Sequence: CVPSPASQGDLHTKPLGTDD

Specifications

Antigen NLRP1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NLRP1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene NLRP1
Gene Alias CARD7/CLR17.1/DEFCAP/DEFCAP-L/S/DKFZp586O1822/KIAA0926/NAC/NALP1/PP1044/SLEV1/VAMAS1
Gene Symbols NLRP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1058-1077 of human NLRP1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22861
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.