Learn More
NLRP4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NLRP4.
Supplier: Abnova Corporation PAB0268
Description
NALPs are cytoplasmic proteins that form a subfamily within the larger CATERPILLER protein family. Most short NALPs, such as NALP4, have an N-terminal pyrin (MEFV; MIM 608107) domain (PYD), followed by a NACHT domain, a NACHT-associated domain (NAD), and a C-terminal leucine-rich repeat (LRR) region. The long NALP, NALP1 (MIM 606636), also has a C-terminal extension containing a function to find domain (FIIND) and a caspase recruitment domain (CARD). NALPs are implicated in the activation of proinflammatory caspases (e.g., CASP1; MIM 147678) via their involvement in multiprotein complexes called inflammasomes (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM
Sequence: FAPKETGKQPRTVIIQGPQSpecifications
NLRP4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NLRP4. | |
In serum (0.05% sodium azide) | |
CLR19.5/FLJ32126/NALP4/PAN2/PYPAF4/RNH2 | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
NLRP4 | |
NLRP4 | |
A synthetic peptide corresponding to amino acids 139-157 of human NLRP4. | |
RUO | |
147945 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only