Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NME1-NME2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128974 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-974 Supplier Abnova Corporation Supplier No. PAB27463
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant NME1-NME2.

The NME1-NME2 mRNA is a naturally occurring co-transcribed product of the neighboring NME1 and NME2 genes. The significance of this co-transcribed mRNA and the function of its predicted protein product have not yet been determined. Alternative splicing of this gene results in different transcript variants encoding distinct isoforms, but the full-length nature of each variant has not been defined. [provided by RefSeq

Sequence: MVLLSTLGIVFQGEGPPISSCDTGTMANCERTFIAIKPDGVQRGLVGEIIKRFEQKGFRLVGLKFMQ

Specifications

Antigen NME1-NME2
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant NME1-NME2.
Dilution Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide)
Gene NME1-NME2
Gene Alias NME2
Gene Symbols NME1-NME2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human NME1-NME2.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 654364
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.