Learn More
NME2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NME2.
Supplier: Abnova Corporation PAB3291
Description
Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq]
Specifications
NME2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NME2. | |
In PBS (0.09% sodium azide) | |
MGC111212/NDPK-B/NDPKB/NM23-H2/NM23B/puf | |
Rabbit | |
Protein G purification | |
RUO | |
4831 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
NME2 | |
NME2 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human NME2. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only