Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOS2, Rabbit, Clone: K13-A, Abnova™

Catalog No. 89110528 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-528 Supplier Abnova Corporation Supplier No. MAB1627
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of NOS2.

Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. This gene encodes a nitric oxide synthase which is expressed in liver and is inducible by a combination of lipopolysaccharide and certain cytokines. Three related pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq

Specifications

Antigen NOS2
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K13-A
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of NOS2.
Dilution Western Blot (1:2000) ELISA (1:20000-1:50000) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide)
Gene NOS2
Gene Alias HEP-NOS/INOS/NOS/NOS2A
Gene Symbols NOS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human NOS2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4843
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.