Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHS1, Rabbit, Clone: Y17-R, Abnova™

Catalog No. 89110537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-537 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-537 Supplier Abnova Corporation Supplier No. MAB1638
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against synthetic peptide of NPHS1.

Nephrin is a kidney glomerular filtration barrier protein that is an essential component of the interpodocyte-spanning slit diaphragm. Mutations in the nephrin gene are associated with congenital nephrotic syndrome (NPHS1; MIM 256300).[supplied by OMIM

Specifications

Antigen NPHS1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Y17-R
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of NPHS1.
Dilution Western Blot (1:5000) ELISA (1:500001:100000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) The optimal working dilution should be determined by the end user.
Formulation In 20mM Tris-HCl, pH 8.0 (10mg/mL BSA, 0.05% sodium azide)
Gene NPHS1
Gene Alias CNF/NPHN
Gene Symbols NPHS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to fifth Ig-like domain (between residues Cys465-Cys528) of human NPHS1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4868
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.