Learn More
NPR1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NPR1.
Supplier: Abnova Corporation PAB3327
Description
Guanylyl cyclases, catalyzing the production of cGMP from GTP, are classified as soluble and membrane forms (Garbers and Lowe, 1994 [PubMed 7982997]). The membrane guanylyl cyclases, often termed guanylyl cyclases A through F, form a family of cell-surface receptors with a similar topographic structure: an extracellular ligand-binding domain, a single membrane-spanning domain, and an intracellular region that contains a protein kinase-like domain and a cyclase catalytic domain. GC-A and GC-B function as receptors for natriuretic peptides; they are also referred to as atrial natriuretic peptide receptor A (NPR1) and type B (NPR2; MIM 108961). Also see NPR3 (MIM 108962), which encodes a protein with only the ligand-binding transmembrane and 37-amino acid cytoplasmic domains. NPR1 is a membrane-bound guanylate cyclase that serves as the receptor for both atrial and brain natriuretic peptides (ANP (MIM 108780) and BNP (MIM 600295), respectively).[supplied by OMIM
Specifications
NPR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NPR1. | |
In PBS (0.09% sodium azide) | |
ANPRA/ANPa/GUC2A/GUCY2A/NPRA | |
Rabbit | |
Protein G purification | |
RUO | |
4881 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) The optimal working dilution should be determined by the end user. | |
NPR1 | |
NPR1 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human NPR1. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only