Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPSR1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89128215 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-215 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-215 Supplier Abnova Corporation Supplier No. PAB26576
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NPSR1.

This gene is a member of the G protein-coupled receptor 1 family and encodes a plasma membrane protein. Increased expression of this gene in ciliated cells of the respiratory epithelium and in bronchial smooth muscle cells is associated with asthma. Mutations in this gene have also been associated with this disease. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq

Specifications

Antigen NPSR1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NPSR1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene NPSR1
Gene Alias ASRT2/GPR154/GPRA/NPSR/PGR14/VRR1
Gene Symbols NPSR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids from 2nd extracellular domain of human NPSR1.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387129
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Hamster, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.