Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPTN, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89129675 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-675 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-675 Supplier Abnova Corporation Supplier No. PAB28168
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NPTN.

This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Specifications

Antigen NPTN
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NPTN.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-20 ug/ml)
Formulation In PBS (0.09% sodium azide)
Gene NPTN
Gene Alias DKFZp686L2477/GP55/GP65/MGC102805/SDFR1/SDR1/np55/np65
Gene Symbols NPTN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 19 amino acids at internal region of human NPTN.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27020
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except ARHGEF3 (60%).
Target Species Bovine, Canine, Equine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.