Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPY Rabbit anti-Human, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic protein of NPY.
Supplier: Abnova Corporation PAB19556
Specifications
NPY | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic protein of NPY. | |
Lyophilized from PBS. | |
PYY4 | |
Rabbit | |
Protein G purification | |
RUO | |
4852 | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
ELISA (1-2 ug/mL) Western Blot (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user. | |
NPY | |
NPY | |
Synthetic protein corresponding to human NPY. | |
100 μg | |
Primary | |
Human, Mouse, Rabbit, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction