Learn More
NR1D2 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NR1D2.
Supplier: Abnova Corporation PAB27800
Description
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described. [provided by RefSeq
Specifications
NR1D2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NR1D2. | |
In PBS (0.09% sodium azide) | |
BD73/EAR-1r/HZF2/Hs.37288/RVR | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
9975 | |
Human, Monkey | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user. | |
NR1D2 | |
NR1D2 | |
A synthetic peptide corresponding to 19 amino acid at ligand-binding domain of human NR1D2. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only