Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1H3/NR1H2 Rabbit anti-Chicken, Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-907 Supplier Abnova Corporation Supplier No. PAB12535
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.

Specifications

Antigen NR1H3/NR1H2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.
Dilution Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Alias LXR-a/LXRA/RLD-1
Gene Symbols NR1H3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 50-150 of human NR1H3/NR1H2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062/7376
Test Specificity This antibody is specific to LXR alpha and beta isoforms.
Target Species Chicken, Human, Mouse, Primate, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.