Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1H3/NR1H2 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89116829 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-829 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-829 Supplier Abnova Corporation Supplier No. PAB13765

Rabbit polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.

Specifications

Antigen NR1H3/NR1H2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NR1H3/NR1H2.
Dilution Western Blot (2-6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.02% sodium azide)
Gene Alias LXR-a/LXRA/RLD-1
Gene Symbols NR1H3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to surrounding amino acids 435 of human NR1H3/NR1H2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062/7376
Test Specificity This antibody detects 64 KDa and 80 KDa proteins representing the alpha and beta subunits of human LXR.
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.