Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nr2c1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118748 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-748 Supplier Abnova Corporation Supplier No. PAB16944
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant Nr2c1.

Specifications

Antigen Nr2c1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Nr2c1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Nr2c1
Gene Alias 4831444H07Rik/80.3/Eenr/TR2/Tr2-11
Gene Symbols Nr2c1
Host Species Rabbit
Immunogen Recombinant protein corresponding to N-terminus of mouse Nr2c1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22025
Test Specificity This antibody has been directly tested for reactivity in mouse, human and rat. specific to the ∼64 KDa TR2 protein in Western blots of testes and nuclear extracts from MEL cell lines.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.