Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR4A1 (phospho S351), Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114270 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-270 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-270 Supplier Abnova Corporation Supplier No. PAB0547
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of NR4A1.

This gene encodes a member of the steroid-thyroid hormone-retinoid receptor superfamily. Expression is induced by phytohemagglutinin in human lymphocytes and by serum stimulation of arrested fibroblasts. The encoded protein acts as a nuclear transcription factor. Translocation of the protein from the nucleus to mitochondria induces apoptosis. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq

Specifications

Antigen NR4A1
Applications Dot Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of NR4A1.
Dilution Dot Blot (1:100-500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene NR4A1
Gene Alias GFRP1/HMR/MGC9485/N10/NAK-1/NGFIB/NP10/NUR77/TR3
Gene Symbols NR4A1
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S351 of human NR4A1.
Purification Method Protein A purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3164
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.