Learn More
NR5A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NR5A1.
Supplier: Abnova Corporation PAB27766
Description
The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect. [provided by RefSeq
Specifications
NR5A1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of NR5A1. | |
In PBS (0.09% sodium azide) | |
AD4BP/ELP/FTZ1/FTZF1/SF-1/SF1 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
2516 | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15-30 ug/mL) The optimal working dilution should be determined by the end user. | |
NR5A1 | |
NR5A1 | |
A synthetic peptide corresponding to 16 amino acid at internal region of human NR5A1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only