Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-NRF-1, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABE1844
Description
Specifically detects NRF-1 in Human, Mouse, Rat samples, and it is validated for Chromatin Immunoprecipitation, Immunocytochemistry, Western Blotting
Nuclear respiratory factor (UniProt Q9WU00; also known as Alpha-pal, Alpha palindromic-binding protein, NRF-1) is encoded by the Nrf1 gene (also known as C87038, D6Ertd415e) in murine species (Gene ID 18181). NRF-1 is a redox-responsive transcription factor that plays an anti-oxidant, pro-survival role in mitochondrial biogenesis process. NRF-1 is upregulated and activated/phosphorylated by Akt in response to oxidant exposure or increased cellular reactive oxygen species (ROS). This results in NRF-1 nuclear translocation, where it binds to DNA as a homodimer and mediates the transcription of mitochondrial transcription factor A (mtTFA) that is needed for mitochondrial DNA transcription.
Specifications
NRF-1 | |
Polyclonal | |
Q9WU00 | |
Rabbit | |
Protein A Purfied | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ChIP Assay, Immunocytochemistry, Western Blot | |
Purified rabbit polyclonal antibody in buffer containing PBS and 0.01% Sodium Azide. | |
Nrf1 | |
KLH-conjugated linear peptide corresponding to mouse NRF-1. | |
200 μg | |
Epigenetics and Nuclear Function | |
NP_001157698 | |
Stable for one year at 2°C to 8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction