Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nrf1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114766 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-766 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-766 Supplier Abnova Corporation Supplier No. PAB10295

Rabbit polyclonal antibody raised against partial recombinant Nrf1.

Specifications

Antigen Nrf1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Nrf1.
Dilution ELISA (1:3000-1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Nrf1
Gene Alias C87038/D6Ertd415e
Gene Symbols Nrf1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-534 of mouse Nrf1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18181
Test Specificity This protein A purified antibody is directed against mouse NRF1.
Target Species Chimpanzee, Frog, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.