Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRP1 (phospho T916), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115973 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-973 Supplier Abnova Corporation Supplier No. PAB12639
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of NRP1.

NRP1 is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor (VEGF; MIM 192240) and semaphorin (see SEMA3A; MIM 603961) family members. NRP1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration, and invasion.[supplied by OMIM

Sequence: LNTQS

Specifications

Antigen NRP1
Applications Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of NRP1.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene NRP1
Gene Alias BDCA4/CD304/DKFZp686A03134/DKFZp781F1414/NP1/NRP/VEGF165R
Gene Symbols NRP1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T916 of human NRP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8829
Test Specificity This antibody recognizes ∼130 KDa of human NRP1 at the phosphorylation site of Thr 916. It does not cross react to non-phospho Neuropilin 1.
Target Species Bovine, Human
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.