Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nrxn1/Nrxn2/Nrxn3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Nrxn1/Nrxn2/Nrxn3.
Supplier: Abnova Corporation PAB8863
Specifications
Nrxn1/Nrxn2/Nrxn3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Nrxn1/Nrxn2/Nrxn3. | |
In PBS (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
Nrxn1 | |
A synthetic peptide corresponding to residues surrounding amino acids 1112 of rat Nrxn3. | |
RUO | |
60391/116595/116508 | |
Bovine, Human, Rat | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
Nrxn1b | |
Rabbit | |
100 μL | |
Primary | |
This antibody recognizes both alpha (∼170 KDa) and beta (∼50 KDa) isoforms of Neurexin3. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction