Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSMAF, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114663 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-663 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-663 Supplier Abnova Corporation Supplier No. PAB10142
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NSMAF.

This gene encodes a WD-repeat protein that binds the cytoplasmic sphingomyelinase activation domain of the 55kDa tumor necrosis factor receptor. This protein is required for TNF-mediated activation of neutral sphingomyelinase and may play a role in regulating TNF-induced cellular responses such as inflammation. Alternative splicing results in multiple transcript variants

Specifications

Antigen NSMAF
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NSMAF.
Dilution Western blot (1 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene NSMAF
Gene Alias FAN
Gene Symbols NSMAF
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human NSMAF.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8439
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.