Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSUN5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89122802 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-122-802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-122-802 Supplier Abnova Corporation Supplier No. PAB21311
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant NSUN5.

This gene encodes a protein with similarity to p120 (NOL1), a 120kDa proliferation-associated nucleolar antigen that is a member of an evolutionarily conserved protein family. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq

Sequence: QLPRFVRVNTLKTCSDDVVDYFKRQGFSYQGRASSLDDLRALKGKHFLLDPLMPELLVFPAQTDLHEHPLYRAGHLIL

Specifications

Antigen NSUN5
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant NSUN5.
Dilution Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene NSUN5
Gene Alias FLJ10267/MGC986/NOL1/NOL1R/NSUN5A/WBSCR20/WBSCR20A/Ynl022cL/p120
Gene Symbols NSUN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human NSUN5.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55695
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.