Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTF3 Rabbit anti-Chicken, Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114357 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-357 Supplier Abnova Corporation Supplier No. PAB0706
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant NTF3.

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse. [provided by RefSeq

Specifications

Antigen NTF3
Applications ELISA, Immunohistochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant NTF3.
Dilution Immunohistochemistry (1:500-1:1000) ELISA (1:500-1:1000) Western Blot (1:500-1:1000) Inhibition assay (1:10-50 in vitro 2-10 ul/g body weight for in vivo) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene NTF3
Gene Alias HDNF/MGC129711/NGF-2/NGF2/NT3
Gene Symbols NTF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human NTF3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4908
Test Specificity A cross reactivity of less than 1% to mouse NGF, recombinant human BDNF and 5% to NT4/5 has been shown by dot blot.
Target Species Chicken, Human, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.