Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTHL1 Rabbit anti-Bovine, Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89115590 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-590 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-590 Supplier Abnova Corporation Supplier No. PAB12089
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of NTHL1.

The protein encoded by this gene is a DNA N-glycosylase of the endonuclease III family. Like a similar protein in E. coli, the encoded protein has DNA glycosylase activity on DNA substrates containing oxidized pyrimidine residues and has apurinic/apyrimidinic lyase activity. [provided by RefSeq

Specifications

Antigen NTHL1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NTHL1.
Dilution Immunocytochemistry (1:800) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NTHL1
Gene Alias NTH1/OCTS3
Gene Symbols NTHL1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human NTHL1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4913
Test Specificity This antibody is specific to NTH1.
Target Species Bovine, Human, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.