Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-NTRK1, Polyclonal, Abnova™ (phospho Y676), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-309 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-309 Supplier Abnova Corporation Supplier No. PAB0628

Specifications

Antigen NTRK1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide)
Gene Symbols NTRK1
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y676 of human NTRK1.
Purification Method Protein G purification
Quantity 400 μL
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.