Learn More
NTRK1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NTRK1.
Supplier: Abnova Corporation PAB24825
Description
This gene encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of this kinase leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in this gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. [provided by RefSeq
Specifications
NTRK1 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
NTRK1 | |
NTRK1 | |
A synthetic peptide corresponding to NTRK1. | |
100 μL | |
Primary | |
NTRK1 polyclonal antibody detects endogenous levels of NTRK1 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of NTRK1. | |
In 1X PBS, pH 7.2 (0.05% sodium azide) | |
DKFZp781I14186/MTC/TRK/TRK1/TRKA/p140-TrkA | |
Rabbit | |
Antigen affinity purification | |
RUO | |
4914 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only