Learn More
NTRK2 (phospho Y509), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of NTRK2.
Supplier: Abnova Corporation PAB24725
Description
This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
NTRK2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of NTRK2. | |
In PBS, pH 7.2 (0.09% sodium azide) | |
GP145-TrkB/TRKB | |
Rabbit | |
Antigen affinity purification | |
RUO | |
4915 | |
Human | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
NTRK2 | |
NTRK2 | |
Synthetic phosphopeptide corresponding to residues surrounding Y509 of NTRK2. | |
100 μL | |
Primary | |
NTRK2 (phospho Y509) polyclonal antibody detects endogenous levels of NTRK2 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only