Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTRK2 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89117005 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-005 Supplier Abnova Corporation Supplier No. PAB14331
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial Ntrk2.

Specifications

Antigen NTRK2
Applications ELISA, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial Ntrk2.
Dilution ELISA (1:10000) Immunohistochemistry (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Ntrk2
Gene Alias AI848316/C030027L06Rik/Tkrb/trkB
Gene Symbols Ntrk2
Host Species Rabbit
Immunogen Glycosylated mouse extracellular domain Ntrk2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18212
Test Specificity Specificity was demonstrated by immunohistochemistry. This antibody was used to stain cryostat sections of the rat peripheral sensory ganglia.
Target Species Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.