Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-NTT4/SLC6A17, Polyclonal,

Catalog No. ABN1733 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1733 Supplier MilliporeSigma™ Supplier No. ABN1733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects NTT4/SLC6A17 in Rat samples, and it is validated for Immunofluorescence, Western Blotting

Sodium-dependent neutral amino acid transporter SLC6A17 (UniProt P31662; also known as B0AT3, Sodium-dependent neurotransmitter transporter NTT4, Solute carrier family 6 member 17) is encoded by the Slc6a17 (also known as Ntt4, Rxt1) gene (Gene ID 613226) in rat species. SLC6A17/B0AT3 is a member of the SLC6 family of proteins, many of which function as presynaptic reuptake transporters. SLC6 transporters are calssified as secondary active transporters because they are driven by electrochemical Na+ or H+ gradients. Na+ gradients are maintained over the plasma membrane of all cells, while H+ gradients are maintained over internal membranes such as synaptic vesicles. The SLC6 members are divided based on their substrates into four subgroups termed GABA, monoamine, amino acid, and neutral amino acids (originally called orphans). SLC6A17/B0AT3 acts as a transporter of a broad range of neutral amino acids with affinity for proline, leucine, glycine, alanine, and glutamine. SLC6A17/B0AT3 is found exclusively in axon terminals of most glutamatergic neurons and in a sub-population of GABAergic neurons throughout the central nervous system (CNS). Although yet to be verified, SLC6A17/B0AT3 could play a role in synaptic remodeling, a process important for the long-term action of antidepressant drugs as well as for other CNS functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTT4/SLC6A17
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal antibody serum with 0.05% sodium azide.
Gene Accession No. P31662
Gene Symbols Slc6a17; Ntt4; Rxt1
Host Species Rabbit
Immunogen GST-tagged recombinant rat Ntt4/Slc6a17 C-terminal cytoplasmic tail.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001028251
Target Species Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.