Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUCB2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89119620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-620 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-620 Supplier Abnova Corporation Supplier No. PAB18034

Rabbit polyclonal antibody raised against synthetic peptide of NUCB2.

Nucleobindin-2 is a calcium-binding EF-hand protein.[supplied by OMIM

Specifications

Antigen NUCB2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of NUCB2.
Dilution ELISA (10 ng/mL) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene NUCB2
Gene Alias NEFA
Gene Symbols NUCB2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus amino acids of human NUCB2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4925
Test Specificity The rabbit anti-human NESFATIN-1 antibody cross reactivity with mouse and rat are expected from sequence similarity.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.