Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUMA1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115588 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-588 Supplier Abnova Corporation Supplier No. PAB12087
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant NUMA1.

Specifications

Antigen NUMA1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant NUMA1.
Dilution Immunofluorescence (1:500) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Gene NUMA1
Gene Alias NUMA
Gene Symbols NUMA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human NUMA1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4926
Test Specificity This antibody is specific to NuMA.
Target Species Human, Mammalia, Mouse, Primate
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.