Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUP155 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89124859 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-124-859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-124-859 Supplier Abnova Corporation Supplier No. PAB23160

Rabbit polyclonal antibody raised against recombinant NUP155.

Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein encoded by this gene does not contain the typical FG repeat sequences found in most vertebrate nucleoporins. Two protein isoforms are encoded by transcript variants of this gene. [provided by RefSeq

Sequence: SKGILLMAASENEDNDILWCVNHDTFPFQKPMMETQMTAGVDGHSWALSAIDELKVDKIITPLNKDHIPITDSPVVVQQHMLPPKKFVLLSAQGS

Specifications

Antigen NUP155
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description nucleoporin 155kDa
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene NUP155
Gene Alias KIAA0791, N155
Gene Symbols NUP155
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human NUP155.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9631
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.