Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUPR1 Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114529 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-529 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-529 Supplier Abnova Corporation Supplier No. PAB0975

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against synthetic peptide of NUPR1.

Specifications

Antigen NUPR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description nuclear protein 1
Formulation In PBS (0.02% sodium azide)
Gene NUPR1
Gene Alias COM1, P8
Gene Symbols NUPR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human NUPR1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26471
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.