Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXF5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130052 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-052 Supplier Abnova Corporation Supplier No. PAB28542
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant NXF5.

This gene is one member of a family of nuclear RNA export factor genes. The encoded protein can bind RNA, and is implicated in mRNA nuclear export. However, this protein has lost several C-terminal protein domains found in other family members that are required for export activity, and may be an evolving pseudogene. Alternatively spliced transcript variants have been described, but most are candidates for nonsense-mediated decay (NMD) and may not express proteins in vivo. [provided by RefSeq

Sequence: TAPYSVKNKLKPGQMEMLKLTMNKRYNVSQQALDLQNLRFDPDLMGRDIDIILNRRNCMAATLKITERNFPELLSLNLCNNKLYQLDGLSDITEKAPKVKTLNLSKNKLESAWELGKVKGLKLEELWLEGNPLCSTFSDQS

Specifications

Antigen NXF5
Applications Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant NXF5.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene NXF5
Gene Symbols NXF5
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human NXF5.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55998
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.