Learn More
NXPH1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NXPH1
Supplier: Abnova Corporation PAB28021
Description
This gene is a member of the neurexophilin family and encodes a secreted protein with a variable N-terminal domain, a highly conserved, N-glycosylated central domain, a short linker region, and a cysteine-rich C-terminal domain. This protein forms a very tight complex with alpha neurexins, a group of proteins that promote adhesion between dendrites and axons. [provided by RefSeq
Sequence: NLTNGGKSELLKSGSSKSTLKHIWTESSKDLSISRLLSQTFRGKENDTDLDLRYDTPEPYSEQDLWDWLRNSTDLQEPRPRAKRRPIVKTGKFKKMFGWSpecifications
NXPH1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant NXPH1 | |
In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
NPH1/Nbla00697 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
30010 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:500 - 1:1000) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
NXPH1 | |
NXPH1 | |
Recombinant protein corresponding to amino acids of human NXPH1 | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only