Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OGFOD2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-960 Supplier Abnova Corporation Supplier No. PAB19425
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of OGFOD2.

Specifications

Antigen OGFOD2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of OGFOD2.
Dilution Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene OGFOD2
Gene Alias DKFZp686H15154/FLJ13491/FLJ37501/MGC120434/MGC120436
Gene Symbols OGFOD2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids near the center region of human OGFOD2.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79676
Test Specificity OGFOD2 antibody is predicted to not cross-react with other TPA1 protein family members. At least three isoforms are known to exist; this antibody will only detect the two larger isoforms.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.