Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLIG2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-744 Supplier Abnova Corporation Supplier No. PAB16940
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Olig2.

Specifications

Antigen OLIG2
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant Olig2.
Dilution Western Blot (1:3000) Immunohistochemistry (1:1000) Immunocytochemistry (1:1000) Immunoprecipitation (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In 200mM Tris, 100mM glycine, pH 8.0
Gene Olig2
Gene Alias AI604895/Bhlhb1/Olg-2/Oligo2/RK17
Gene Symbols Olig2
Host Species Rabbit
Immunogen Recombinant protein corresponding to mouse Olig2.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50913
Test Specificity This antibody has been directly tested for reactivity in human, mouse and rat. specific to the ∼32 KDa Olig2 protein in Western blots. This antibody also works well for immunohistochemistry and immunocytochemistry.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.