Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPRD1 Rabbit anti-Dog, Horse, Human, Monkey, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OPRD1.
Supplier: Abnova Corporation PAB26244
Specifications
OPRD1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of OPRD1. | |
In PBS (0.09% sodium azide) | |
OPRD | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
4985 | |
Canine, Equine, Human, Monkey, Rabbit | |
Liquid |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
OPRD1 | |
OPRD1 | |
A synthetic peptide corresponding to 18 amino acid at extracellular domain of human OPRD1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction