Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
orf1ab (SARS orf1ab) Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant SARS orf1ab.
Supplier: Abnova Corporation PAB11372
Specifications
| orf1ab (SARS orf1ab) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant SARS orf1ab. | |
| In antiserum (0.01% sodium azide) | |
| orf1ab | |
| Recombinant His fusion protein corresponding to full length SARS orf1ab. | |
| RUO | |
| 1489680 | |
| Virus | |
| Liquid |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunofluorescence (1:300) Immunoprecipitation (1:60) Immunoelectron Microscopy (1:100) The optimal working dilution should be determined by the end user. | |
| orf1ab | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is directed against SARS-Coronavirus nsp8 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction