Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Os08g0103300, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89120667 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-667 Supplier Abnova Corporation Supplier No. PAB19135
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Os08g0103300.

Specifications

Antigen Os08g0103300
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Os08g0103300.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Os08g0103300
Gene Symbols Os08g0103300
Host Species Rabbit
Immunogen A synthetic peptide corresponding to Oryza sativa Os08g0103300.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4344429
Test Specificity It recognizes HSP in rice tissue lysate.
Target Species Rice
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.