Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OS9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115584 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-584 Supplier Abnova Corporation Supplier No. PAB12082
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of OS9.

This gene encodes a protein that is highly expressed in osteosarcomas. This protein binds to the hypoxia-inducible factor 1 (HIF-1), a key regulator of the hypoxic response and angiogenesis, and promotes the degradation of one of its subunits. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Specifications

Antigen OS9
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of OS9.
Dilution Immunofluorescence (0.2-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.01% sodium azide)
Gene OS9
Gene Symbols OS9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 300-400 of human OS9.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10956
Test Specificity This antibody is specific to isoforms 1 and 2 of human OS-9.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.