Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTOA, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-507 Supplier Abnova Corporation Supplier No. PAB23799
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant OTOA.

The protein encoded by this gene is specifically expressed in the inner ear, and is located at the interface between the apical surface of the inner ear sensory epithelia and their overlying acellular gels. It is prposed that this protein is involved in the attachment of the inner ear acellular gels to the apical surface of the underlying nonsensory cells. Mutations in this gene are associated with autosomal recessive deafness type 22 (DFNB22). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: RCMEEDTFIRTVELLGAVQGFSRPQLMTLKEKAIQVWDMPSYWREHHIVSLGRIALALNESELEQLDLSSIDTVASLSWQTEWTP

Specifications

Antigen OTOA
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant OTOA.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene OTOA
Gene Alias DFNB22/FLJ32773/MGC157747/MGC39813
Gene Symbols OTOA
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human OTOA.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146183
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.