Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2RY6 Rabbit anti-Bovine, Hamster, Human, Monkey, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89128038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-038 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-038 Supplier Abnova Corporation Supplier No. PAB26397
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of P2RY6.

The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to UDP, partially responsive to UTP and ADP, and not responsive to ATP. Four transcript variants encoding the same isoform have been identified for this gene. [provided by RefSeq

Specifications

Antigen P2RY6
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of P2RY6.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (17-34 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene P2RY6
Gene Alias MGC15335/P2Y6
Gene Symbols P2RY6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at 2nd cytoplasmic domain of human P2RY6.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5031
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except MOGAT1 (47%).
Target Species Bovine, Hamster, Human, Monkey, Pig, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.