Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-p47-phox, Polyclonal,

Catalog No. ABN1480MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1480MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1480MI Supplier MilliporeSigma™ Supplier No. ABN1480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects p47-phox in Human, Mouse samples, and it is validated for Immunohistochemistry, Western Blotting

Neutrophil cytosol factor 1 (UniProt Q09014), also known as 47kDa neutrophil oxidase factor, NADPH oxidase subunit, Ncf-1, NCF-47K, Neutrophil NADPH oxidase factor 1, p47-phox, p47phox) is encoded by the Ncf1 gene in murine species. Activation of NADPH-oxidase dependent superoxide generation requires the participation of several cytosolic factors, including p47-phox, p67-phox, p40-phox and Rac. In the resting cell, p47phox, p67phox, and p40phox form a large molecular weight cytosolic complex. Activation of the NADPH oxidase is initiated by the assembly of p47phox, p67phox, and Rac with cytochrome b558 in a 1:1:1:1 complex in the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p47-phox
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q09014
Gene Symbols Ncf1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of mouse p47-phox.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001272966
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.