Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P4hb, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-608 Supplier Abnova Corporation Supplier No. PAB15416
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of P4hb.

Specifications

Antigen P4hb
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of P4hb.
Dilution Western Blot (1:4000) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene P4hb
Gene Alias PDIR
Gene Symbols P4hb
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to rat P4hb.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25506
Test Specificity Detects ∼58KDa.
Target Species Bovine, Canine, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.