Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P4HTM, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115891 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-891 Supplier Abnova Corporation Supplier No. PAB12516
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant P4HTM.

The product of this gene belongs to the family of prolyl 4-hydroxylases. This protein is a prolyl hydroxylase that may be involved in the degradation of hypoxia-inducible transcription factors under normoxia. It plays a role in adaptation to hypoxia and may be related to cellular oxygen sensing. Alternatively spliced variants encoding different isoforms have been identified. [provided by RefSeq

Specifications

Antigen P4HTM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant P4HTM.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene P4HTM
Gene Alias EGLN4/HIFPH4/P4H-TM/PH-4/PH4/PHD4
Gene Symbols P4HTM
Host Species Rabbit
Immunogen Recombinant protein corresponding to C-terminus of human P4HTM.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54681
Test Specificity This antibody is specific to prolyl hydroxylase 4.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.