Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAF1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-930 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-930 Supplier Abnova Corporation Supplier No. PAB14100

Rabbit polyclonal antibody raised against full-length recombinant PAF1.

PAF1, parafibromin (CDC73; MIM 607393), LEO1 (MIM 610507), and CTR9 (MIM 609366) form the PAF protein complex that associates with the RNA polymerase II subunit POLR2A (MIM 180660) and with a histone methyltransferase complex (Rozenblatt-Rosen et al., 2005 [PubMed 15632063]). The PAF complex also has a role in histone monoubiquitination (Zhu et al., 2005 [PubMed 16307923]).[supplied by OMIM

Specifications

Antigen PAF1
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant PAF1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide, 0.05% proclin 300)
Gene PAF1
Gene Alias F23149_1/FLJ11123/PD2
Gene Symbols PAF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human PAF1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54623
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.